missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Iodo-1,2,4,5-tetramethylbenzene 98.0+%, TCI America™
$61.51 - $206.90
Chemical Identifiers
| CAS | 2100-25-6 |
|---|---|
| Molecular Formula | C10H13I |
| Molecular Weight (g/mol) | 260.12 |
| MDL Number | MFCD00013705 |
| InChI Key | MCRSYYLHVQGFNR-UHFFFAOYSA-N |
| Synonym | 2,3,5,6-tetramethyliodobenzene, iododurene, 3-iododurene, benzene, 3-iodo-1,2,4,5-tetramethyl, 1-iodo-2,3,5,6-tetramethylbenzene, 3-iodo-1,2,4,5-tetramethyl-benzene, acmc-209fgl, maybridge1_002808, 3-iodo-1,4,5-tetramethylbenzene, iodo-2,3,5,6-tetramethylbenzene |
| PubChem CID | 75011 |
| IUPAC Name | 3-iodo-1,2,4,5-tetramethylbenzene |
| SMILES | CC1=CC(C)=C(C)C(I)=C1C |
Chemical Identifiers
| 2100-25-6 | |
| 260.12 | |
| MCRSYYLHVQGFNR-UHFFFAOYSA-N | |
| 75011 | |
| CC1=CC(C)=C(C)C(I)=C1C |
| C10H13I | |
| MFCD00013705 | |
| 2,3,5,6-tetramethyliodobenzene, iododurene, 3-iododurene, benzene, 3-iodo-1,2,4,5-tetramethyl, 1-iodo-2,3,5,6-tetramethylbenzene, 3-iodo-1,2,4,5-tetramethyl-benzene, acmc-209fgl, maybridge1_002808, 3-iodo-1,4,5-tetramethylbenzene, iodo-2,3,5,6-tetramethylbenzene | |
| 3-iodo-1,2,4,5-tetramethylbenzene |
Specifications
| 2100-25-6 | |
| White-Yellow | |
| C10H13I | |
| 1 g | |
| MCRSYYLHVQGFNR-UHFFFAOYSA-N | |
| 3-iodo-1,2,4,5-tetramethylbenzene | |
| 75011 | |
| ≥98.0% (GC) | |
| 3-Iodo-1,2,4,5-tetramethylbenzene |
| 80°C | |
| 290°C | |
| MFCD00013705 | |
| 2,3,5,6-tetramethyliodobenzene, iododurene, 3-iododurene, benzene, 3-iodo-1,2,4,5-tetramethyl, 1-iodo-2,3,5,6-tetramethylbenzene, 3-iodo-1,2,4,5-tetramethyl-benzene, acmc-209fgl, maybridge1_002808, 3-iodo-1,4,5-tetramethylbenzene, iodo-2,3,5,6-tetramethylbenzene | |
| CC1=CC(C)=C(C)C(I)=C1C | |
| 260.12 | |
| 260.12 | |
| Crystalline Powder |
Safety and Handling
TSCA : No