accessibility menu, dialog, popup

UserName

NClFOH

CAS RN 209328-72-3

IUPAC Name: 5-chloro-6-fluoropyridin-3-ol
Synonyms: 5-chloro-6-fluoropyridin-3-ol
Molecular Weight (g/mol): 147.53
Molecular Formula: C5H3ClFNO
InChi Key: IETLNKVWDZYCPN-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=C(F)N=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results