accessibility menu, dialog, popup

UserName

CAS RN 2090-05-3

IUPAC Name: calcium dibenzoate
Synonyms: calcium benzoate
Molecular Weight (g/mol): 282.31
Molecular Formula: C14H10CaO4
InChi Key: NSQPPSOSXWOZNH-UHFFFAOYSA-L
SMILES: [Ca++].[O-]C(=O)C1=CC=CC=C1.[O-]C(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)