accessibility menu, dialog, popup

UserName

CAS RN 2083627-02-3

CH3SOONNNNHNO

CAS RN 2083627-02-3

IUPAC Name: N-[(furan-2-yl)methyl]-8-(4-methanesulfonylphenyl)-[1,2,4]triazolo[4,3-c]pyrimidin-5-amine
Synonyms: N-(furan-2-ylmethyl)-8-(4-methanesulfonylphenyl)-[1,2,4]triazolo[4,3-c]pyrimidin-5-amine
Molecular Weight (g/mol): 369.4
Molecular Formula: C17H15N5O3S
InChi Key: DYIRSNMPIZZNBK-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)C1=CN=C(NCC2=CC=CO2)N2C=NN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results