missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EED226, 95%

Catalog No. AC468141000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468141000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC468141000 Supplier Thermo Scientific™ Supplier No. 468141000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material EED226
CAS 2083627-02-3
Synonym N-(2-Furanylmethyl)-8-[4-(methylsulfonyl)phenyl]-1,2,4-triazolo[4,3-c]pyrimidin-5-amine
Quantity 100 mg
Molecular Formula C17 H15 N5 O3 S
Formula Weight 369.40
Physical Form Solid
Product Type Reagent
Purity 95%