CAS RN 20824-56-0
IUPAC Name:
diammonium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate
Synonyms:
diammonium edta
Molecular Weight (g/mol):
326.31
Molecular Formula:
C10H22N4O8
InChi Key:
KYQODXQIAJFKPH-UHFFFAOYSA-N
SMILES:
[NH4+].[NH4+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
1
–
1
of
1
results
1
Ethylenediaminetetraacetic acid diammonium salt hydrate
CAS: 20824-56-0 Molecular Formula: C10H24N4O9 Molecular Weight (g/mol): 344.321 MDL Number: MFCD00150463 InChI Key: WRSUMHYBOYWMTD-UHFFFAOYSA-N Synonym: ethylenediaminetetraacetic acid diammonium salt hydrate,edetic acid diamine hydrate,diazanium,2-2-bis carboxymethyl amino ethyl-carboxylatome,ammonium 2,2-2-bis carboxymethyl amino ethyl azanediyl diacetate hydrate PubChem CID: 71311375 IUPAC Name: azane;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;hydrate SMILES: C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.N.N.O