accessibility menu, dialog, popup

UserName

CAS RN 207974-07-0

H3CSFFFNH2

CAS RN 207974-07-0

IUPAC Name: 2-(methylsulfanyl)-5-(trifluoromethyl)aniline
Synonyms: 2-(methylsulfanyl)-5-(trifluoromethyl)aniline
Molecular Weight (g/mol): 207.21
Molecular Formula: C8H8F3NS
InChi Key: OQFOLJGOVFYDHZ-UHFFFAOYSA-N
SMILES: CSC1=C(N)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results