CAS RN 207234-02-4
IUPAC Name:
1-dodecylpyridin-1-ium chloride
Synonyms:
laurylpyridinium chloride
Molecular Weight (g/mol):
283.88
Molecular Formula:
C17H30ClN
InChi Key:
GKQHIYSTBXDYNQ-UHFFFAOYSA-M
SMILES:
[Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1
1
–
1
of
1
results
1
n-Dodecylpyridinium chloride hydrate, 98%
CAS: 207234-02-4 Molecular Formula: C17H30ClN Molecular Weight (g/mol): 283.88 MDL Number: MFCD00011987 InChI Key: GKQHIYSTBXDYNQ-UHFFFAOYSA-M Synonym: 1-dodecylpyridin-1-ium chloride hydrate,laurylpyridinum chloride,1-dodecylpyridinium chloride hydrate,1-dodecylpyridin-1-ium hydrate chloride,c17h30n.cl.h2o,n-dodecylpyridinium chloride hydrate,dodecylpyridinium chloride monohydrate,n-dodecylpyridinium chloride monohydrate,1-dodecylpyridin-1-ium chloride-water 1/1/1 PubChem CID: 16211802 IUPAC Name: 1-dodecylpyridin-1-ium;chloride;hydrate SMILES: [Cl-].CCCCCCCCCCCC[N+]1=CC=CC=C1