accessibility menu, dialog, popup

UserName

CAS RN 206559-36-6

IUPAC Name: 1-(benzyloxy)-3-isothiocyanatobenzene
Synonyms: 1-(benzyloxy)-3-isothiocyanatobenzene
Molecular Weight (g/mol): 241.31
Molecular Formula: C14H11NOS
InChi Key: QJLFNDXMZQJMDL-UHFFFAOYSA-N
SMILES: S=C=NC1=CC(OCC2=CC=CC=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results