accessibility menu, dialog, popup

UserName

CAS RN 205319-06-8

IUPAC Name: dichloropalladiumbis(ylium); {2-[2-(diphenylphosphanyl)phenoxy]phenyl}diphenylphosphane
Synonyms: dichloropalladiumbis(ylium); dpephos
Molecular Weight (g/mol): 715.89
Molecular Formula: C36H28Cl2OP2Pd
InChi Key: QZIOEBWDZNUTLZ-UHFFFAOYSA-L
SMILES: Cl[Pd++]Cl.O(C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results