accessibility menu, dialog, popup

UserName

CAS RN 2052-01-9

IUPAC Name: 2-bromo-2-methylpropanoic acid
Synonyms: 2-bromo-2-methylpropanoic acid
Molecular Weight (g/mol): 167
Molecular Formula: C4H7BrO2
InChi Key: XXSPGBOGLXKMDU-UHFFFAOYSA-N
SMILES: CC(C)(Br)C(O)=O
Boiling Point 
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (6)
  • (1)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (3)
  • (2)
  • (2)
Physical Form 
  • (3)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (4)
  • (3)
  • (6)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)