missing translation for 'onlineSavingsMsg'
Learn More

2-Bromoisobutyric acid, 98%

Catalog No. AAA1585436
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1585436 500 g
AAA1585414 25 g
AAA1585422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1585436 Supplier Thermo Scientific Chemicals Supplier No. A1585436
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2052-01-9 | C4H7BrO2 | 167.00 g/mol

2-Bromoisobutyric acid is used in the synthesis of dextran macroinitiator for atom transfer radical polymerization (ATRP) by partial esterification of hydroxyl group of the polysaccharide. The multifunctional silica colloids were prepared by coating them with 2-bromo-2-methylpropionic acid stabilized quantum dots.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoisobutyric acid is used in the synthesis of dextran macroinitiator for atom transfer radical polymerization (ATRP) by partial esterification of hydroxyl group of the polysaccharide. The multifunctional silica colloids were prepared by coating them with 2-bromo-2-methylpropionic acid stabilized quantum dots.

Solubility
Soluble in alcohol and ether. Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases.

Chemical Identifiers

CAS 2052-01-9
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.00
MDL Number MFCD00004168
InChI Key XXSPGBOGLXKMDU-UHFFFAOYSA-N
Synonym 2-bromoisobutyric acid, 2-bromo-2-methylpropionic acid, isobromobutyric acid, propanoic acid, 2-bromo-2-methyl, alpha-bromoisobutyric acid, .alpha.-bromoisobutyric acid, 2-bromo-2-methyl-propanoic acid, propionic acid, 2-bromo-2-methyl, unii-821p2r6b0a, ccris 9046
PubChem CID 74924
IUPAC Name 2-bromo-2-methylpropanoic acid
SMILES CC(C)(C(=O)O)Br

Specifications

Melting Point 42°C to 46°C
Boiling Point 198°C to 200°C
Flash Point >110°C (230°F)
Quantity 500 g
UN Number UN3261
Beilstein 1745543
Merck Index 14,1421
Solubility Information Soluble in alcohol and ether. Slightly soluble in water.
Formula Weight 167
Percent Purity 98%
Chemical Name or Material 2-Bromoisobutyric acid
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318-H317
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 218-139-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only