missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-2-methylpropionic acid, 98%

Catalog No. AC305201000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305201000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC305201000 Supplier Thermo Scientific Chemicals Supplier No. 305201000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2052-01-9 | C4H7BrO2 | 167.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2052-01-9
Molecular Formula C4H7BrO2
Molecular Weight (g/mol) 167.01
MDL Number MFCD00004168
InChI Key XXSPGBOGLXKMDU-UHFFFAOYSA-N
Synonym α-Bromoisobutyric acid
IUPAC Name 2-bromo-2-methylpropanoic acid
SMILES CC(C)(Br)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 41°C to 46°C
Color Colorless to Yellow
Boiling Point 198°C to 200°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula BrC(CH3)2CO2H
Quantity 100 g
Merck Index 15, 1431
Solubility Information Solubility in water: slightly soluble
Formula Weight 167.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Bromo-2-methylpropionic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 218-139-2

RUO – Research Use Only