CAS RN 20469-61-8
CAS RN 20469-61-8
IUPAC Name:
1-methoxy-2,3,5-trimethylbenzene
Synonyms:
1-methoxy-2,3,5-trimethylbenzene
Molecular Weight (g/mol):
150.22
Molecular Formula:
C10H14O
InChi Key:
AWONIZVBKXHWJP-UHFFFAOYSA-N
SMILES:
COC1=CC(C)=CC(C)=C1C
1
–
1
of
1
results
1
2,3,5-Trimethylanisole 97.0+%, TCI America™
CAS: 20469-61-8 Molecular Formula: C10H14O Molecular Weight (g/mol): 150.22 MDL Number: MFCD00142783 InChI Key: AWONIZVBKXHWJP-UHFFFAOYSA-N Synonym: 2,3,5-trimethylanisole,anisole, 2,3,5-trimethyl,benzene, 1-methoxy-2,3,5-trimethyl,1-methoxy-2,3,5-trimethyl-benzene,pubchem2688,acmc-209fam,2,3,5-trimethylanizole,3-methoxy-1,2,5-trimethylbenzene PubChem CID: 88555 IUPAC Name: 1-methoxy-2,3,5-trimethylbenzene SMILES: COC1=CC(C)=CC(C)=C1C