accessibility menu, dialog, popup

UserName

CAS RN 2043-43-8

H3COHNH2O

CAS RN 2043-43-8

IUPAC Name: 2-hydroxypropanamide
Synonyms: lactamide
Molecular Weight (g/mol): 89.09
Molecular Formula: C3H7NO2
InChi Key: SXQFCVDSOLSHOQ-UHFFFAOYNA-N
SMILES: CC(O)C(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results