CAS RN 203927-98-4
IUPAC Name:
[7-(dihydroxyboranyl)-9,9-dihexyl-9H-fluoren-2-yl]boronic acid
Synonyms:
7-(dihydroxyboranyl)-9,9-dihexylfluoren-2-ylboronic acid
Molecular Weight (g/mol):
422.18
Molecular Formula:
C25H36B2O4
InChi Key:
QFWOJNOZWMHNTK-UHFFFAOYSA-N
SMILES:
CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O
1
–
1
of
1
results
1
Promotions Available
9,9-Di-n-hexylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals
9,9-Di-n-hexylfluorene-2,7-diboronic acid, 97%, Thermo Scientific Chemicals
CAS: 203927-98-4 Molecular Formula: C25H36B2O4 Molecular Weight (g/mol): 422.18 MDL Number: MFCD03427279 InChI Key: QFWOJNOZWMHNTK-UHFFFAOYSA-N Synonym: 9,9-dihexylfluorene-2,7-diboronic acid,9,9-dihexyl-9h-fluorene-2,7-diyl diboronic acid,7-borono-9,9-dihexyl-fluoren-2-yl boronic acid,boronic acid,b,b'-9,9-dihexyl-9h-fluorene-2,7-diyl bis,acmc-20alvw,9,9-dihexyl-fluorene-2,7-diboronic acid,9,9-dihexyl-9h-fluorene-2,7-diboronic acid,7-borono-9,9-dihexylfluoren-2-yl boronic acid,2,7-di dihydroxyboryl-9,9-dihexyl-9h-fluorene PubChem CID: 4524285 SMILES: CCCCCCC1(CCCCCC)C2=C(C=CC(=C2)B(O)O)C2=C1C=C(C=C2)B(O)O