accessibility menu, dialog, popup

UserName

H3CCH3H3COOOHONHOO

CAS RN 203854-49-3

IUPAC Name: 6-(tert-butoxy)-3-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-oxohexanoic acid
Synonyms: 6-(tert-butoxy)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-6-oxohexanoic acid
Molecular Weight (g/mol): 439.51
Molecular Formula: C25H29NO6
InChi Key: XPCDWOCHPTYDPV-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)CCC(CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results