accessibility menu, dialog, popup

UserName

CAS RN 2033-89-8

IUPAC Name: 3,4-dimethoxyphenol
Synonyms: veratryl alcohol
Molecular Weight (g/mol): 154.17
Molecular Formula: C8H10O3
InChi Key: SMFFZOQLHYIRDA-UHFFFAOYSA-N
SMILES: COC1=CC=C(O)C=C1OC
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)