missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenol, 98%

Catalog No. AAA1134422
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1134422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1134422 Supplier Thermo Scientific Chemicals Supplier No. A1134422
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2033-89-8 | C8H10O3 | 154.165 g/mol

3,4-Dimethoxyphenol is used in the preparation of 5,6-dimethoxy benzofuranone derivatives and multi-target anti Alzheimer compounds. It is also used in the preparation of 3,4-dimethoxyphenyl-beta-D-glucopyranoside and 4-(but-2-enyloxy)-1,2-dimethoxybenzene. Further, it serves as a precursor for 4H-chromenes synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethoxyphenol is used in the preparation of 5,6-dimethoxy benzofuranone derivatives and multi-target anti Alzheimer compounds. It is also used in the preparation of 3,4-dimethoxyphenyl-beta-D-glucopyranoside and 4-(but-2-enyloxy)-1,2-dimethoxybenzene. Further, it serves as a precursor for 4H-chromenes synthesis.

Solubility
Soluble in water.

Notes
Light sensitive. Store in a cool place. Incompatible with bases, acid chlorides, acid anhydrides and oxidizing agents.

Chemical Identifiers

CAS 2033-89-8
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
MDL Number MFCD00008390
InChI Key SMFFZOQLHYIRDA-UHFFFAOYSA-N
Synonym phenol, 3,4-dimethoxy, 4-hydroxyveratrole, 3,4-bis methyloxy phenol, unii-38b43wcu83, 3,4-dimethoxy phenol, 3,4-dimethoxyphel, 3,4,dimethoxyphenol, phenol,4-dimethoxy, 3,4-dimethoxy-pheno, pubchem4113
PubChem CID 16251
IUPAC Name 3,4-dimethoxyphenol
SMILES COC1=C(C=C(C=C1)O)OC

Specifications

Melting Point 79°C to 82°C
Boiling Point 167°C (14 mmHg)
Quantity 100 g
Beilstein 1366650
Sensitivity Light sensitive
Solubility Information Soluble in water.
Formula Weight 154.16
Percent Purity 98%
Chemical Name or Material 3,4-Dimethoxyphenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 217-995-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only