missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxyphenol 97.0+%, TCI America™

Catalog No. D32215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32215G 5 g
D322125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D32215G Supplier TCI America Supplier No. D32215G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2033-89-8
Molecular Formula C8H10O3
Molecular Weight (g/mol) 154.165
InChI Key SMFFZOQLHYIRDA-UHFFFAOYSA-N
Synonym phenol, 3,4-dimethoxy, 4-hydroxyveratrole, 3,4-bis methyloxy phenol, unii-38b43wcu83, 3,4-dimethoxy phenol, 3,4-dimethoxyphel, 3,4,dimethoxyphenol, phenol,4-dimethoxy, 3,4-dimethoxy-pheno, pubchem4113
PubChem CID 16251
IUPAC Name 3,4-dimethoxyphenol
SMILES COC1=C(C=C(C=C1)O)OC

Specifications

Melting Point 80°C
Boiling Point 167°C
Quantity 5 g
Formula Weight 154.17
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,4-Dimethoxyphenol