accessibility menu, dialog, popup

UserName

CAS RN 20306-22-3

IUPAC Name: 1,4-bis(prop-2-en-1-yl) cyclohexane-1,4-dicarboxylate
Synonyms: 1,4-bis(prop-2-en-1-yl) cyclohexane-1,4-dicarboxylate
Molecular Weight (g/mol): 252.31
Molecular Formula: C14H20O4
InChi Key: ZXTPJLLIHIDBKQ-UHFFFAOYSA-N
SMILES: C=CCOC(=O)C1CCC(CC1)C(=O)OCC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results