accessibility menu, dialog, popup

UserName

CAS RN 2029-94-9

CH3OOOH3C

CAS RN 2029-94-9

IUPAC Name: 3,4-diethoxybenzaldehyde
Synonyms: 3,4-diethoxybenzaldehyde
Molecular Weight (g/mol): 194.23
Molecular Formula: C11H14O3
InChi Key: SSTRYEXQYQGGAS-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=O)C=C1OCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results