accessibility menu, dialog, popup

UserName

CAS RN 202865-68-7

IUPAC Name: 1-(3-bromo-4-fluorophenyl)methanamine hydrochloride
Synonyms: 1-(3-bromo-4-fluorophenyl)methanamine hydrochloride
Molecular Weight (g/mol): 240.5
Molecular Formula: C7H8BrClFN
InChi Key: RSFNYNQGERWCMT-UHFFFAOYSA-N
SMILES: Cl.NCC1=CC(Br)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results