accessibility menu, dialog, popup

UserName

H3CCH3OO

CAS RN 20279-29-2

IUPAC Name: phenyl 2-methylpropanoate
Synonyms: phenyl 2-methylpropanoate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: PIZOACXKIKXRDJ-UHFFFAOYSA-N
SMILES: CC(C)C(=O)OC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results