accessibility menu, dialog, popup

UserName

CAS RN 202256-86-8

NH2NH2

CAS RN 202256-86-8

IUPAC Name: 4-(2-aminoethyl)cyclohexan-1-amine
Synonyms: 4-(2-aminoethyl)cyclohexan-1-amine
Molecular Weight (g/mol): 142.25
Molecular Formula: C8H18N2
InChi Key: VFGQLNVWQRKCAU-UHFFFAOYSA-N
SMILES: NCCC1CCC(N)CC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results