accessibility menu, dialog, popup

UserName

CAS RN 2010-15-3

ONHSN

CAS RN 2010-15-3

IUPAC Name: 3-phenyl-2-sulfanylideneimidazolidin-4-one
Synonyms: 3-phenyl-2-sulfanylideneimidazolidin-4-one
Molecular Weight (g/mol): 192.24
Molecular Formula: C9H8N2OS
InChi Key: ZZRIQDWDJVLELF-UHFFFAOYSA-N
SMILES: O=C1CNC(=S)N1C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results