accessibility menu, dialog, popup

UserName

CAS RN 20017-68-9

Br

CAS RN 20017-68-9

IUPAC Name: (3-bromo-1-phenylpropyl)benzene
Synonyms: (3-bromo-1-phenylpropyl)benzene
Molecular Weight (g/mol): 275.19
Molecular Formula: C15H15Br
InChi Key: SLHSRCBFPHCSGL-UHFFFAOYSA-N
SMILES: BrCCC(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results