accessibility menu, dialog, popup

UserName

H3COClCl

CAS RN 1984-65-2

IUPAC Name: 1,3-dichloro-2-methoxybenzene
Synonyms: 1,3-dichloro-2-methoxybenzene
Molecular Weight (g/mol): 177.02
Molecular Formula: C7H6Cl2O
InChi Key: KZLMCDNAVVJKPX-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results