accessibility menu, dialog, popup

UserName

NaH3COO

CAS RN 1984-06-1

IUPAC Name: sodium octanoate
Synonyms: sodium caprylate
Molecular Weight (g/mol): 166.2
Molecular Formula: C8H15NaO2
InChi Key: BYKRNSHANADUFY-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCC([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (11)
  • (1)
Percent Purity 
  • (2)
  • (8)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)
  • (8)
Grade 
  • (8)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (13)
  • (8)
  • (8)
  • (2)
  • (5)
  • (3)
  • (11)
  • (1)
  • (2)
  • (8)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (8)
  • (8)