missing translation for 'onlineSavingsMsg'
Learn More

Sodium octanoate, 96%

Catalog No. AAA1345136
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1345136 500 g
AAA1345122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1345136 Supplier Thermo Scientific Chemicals Supplier No. A1345136
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1984-06-1 | C8H15NaO2 | 166.20 g/mol

Sodium octanoate may be used as a protein stabilizer for molecules such as albumin and fibrinogen under conditions such heat sterilization. Also used for blood plasma fractionation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium octanoate may be used as a protein stabilizer for molecules such as albumin and fibrinogen under conditions such heat sterilization. Also used for blood plasma fractionation.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 1984-06-1
MDL Number MFCD00058511

Specifications

Melting Point >300°C
Molecular Formula C8H15NaO2
Quantity 500 g
Beilstein 3597723
Synonym sodium octanoate, sodium caprylate, octanoic acid, sodium salt, sodium n-octanoate, sodium octoate, caprylic acid sodium salt, natrium octanoat, octanoic acid sodium salt, unii-9xtm81vk2b, 9xtm81vk2b
Solubility Information Soluble in water.
InChI Key BYKRNSHANADUFY-UHFFFAOYSA-M
SMILES CCCCCCCC(=O)[O-].[Na+]
IUPAC Name sodium;octanoate
Molecular Weight (g/mol) 166.20
PubChem CID 23664772
Formula Weight 166.2
Percent Purity 96%
Chemical Name or Material Sodium octanoate
Show More Show Less
Hazard Category H315-H319-H335-H373-H500
Precautionary Statement P260-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P314-P332+P313-P362-P501c
EINECSNumber 217-850-5
RTECSNumber RH0787000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only