accessibility menu, dialog, popup

UserName

CH3OHONOO

CAS RN 1975-51-5

IUPAC Name: 2-methyl-4-nitrobenzoic acid
Synonyms: 2-methyl-4-nitrobenzoic acid
Molecular Weight (g/mol): 181.15
Molecular Formula: C8H7NO4
InChi Key: XXXOBNJIIZQSPT-UHFFFAOYSA-N
SMILES: CC1=CC(=CC=C1C(O)=O)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)