accessibility menu, dialog, popup

UserName

CH3H3CCH3Cl

CAS RN 19692-45-6

IUPAC Name: 1-tert-butyl-4-(chloromethyl)benzene
Synonyms: 1-tert-butyl-4-(chloromethyl)benzene
Molecular Weight (g/mol): 182.69
Molecular Formula: C11H15Cl
InChi Key: WAXIFMGAKWIFDQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(CCl)C=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (3)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)