accessibility menu, dialog, popup

UserName

CAS RN 196599-80-1

IUPAC Name: 1,2-bis(4-methoxyphenyl)-2-oxoethyl N-cyclohexylcarbamate
Synonyms: 1,2-bis(4-methoxyphenyl)-2-oxoethyl N-cyclohexylcarbamate
Molecular Weight (g/mol): 397.47
Molecular Formula: C23H27NO5
InChi Key: MTNUOOJIYAKURM-UHFFFAOYNA-N
SMILES: COC1=CC=C(C=C1)C(OC(=O)NC1CCCCC1)C(=O)C1=CC=C(OC)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results