accessibility menu, dialog, popup

UserName

CH3H3CCH3SOON(E/Z)

CAS RN 196309-76-9

IUPAC Name: 3-(4-tert-butylbenzenesulfonyl)prop-2-enenitrile
Synonyms: 3-(4-tert-butylbenzenesulfonyl)prop-2-enenitrile
Molecular Weight (g/mol): 249.33
Molecular Formula: C13H15NO2S
InChi Key: VHKZGNPOHPFPER-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)C=CC#N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Product Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results