missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals BAY 11-7085, 99%

Catalog No. AAJ67222MA
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67222MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ67222MA Supplier Thermo Scientific Chemicals Supplier No. J67222MA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 196309-76-9 | C13H15NO2S, C13H15NO2S | 249.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 196309-76-9
Molecular Formula C13H15NO2S
Synonym (E)-3-(4-tert-Butylphenylsulfonyl)acrylonitrile;

Specifications

Quantity 10 mg
Description White
Formula Weight 249.33
Sensitivity Light Sensitive
Melting Point 80°C to 82°C
Product Type BAY 11-7085, 99%
Form Solid
Assay 99%

RUO – Research Use Only