CAS RN 19617-44-8
IUPAC Name:
ethyl N-(ethoxycarbonyl)carbamate
Synonyms:
ethyl N-(ethoxycarbonyl)carbamate
Molecular Weight (g/mol):
161.16
Molecular Formula:
C6H11NO4
InChi Key:
PQVSTLUFSYVLTO-UHFFFAOYSA-N
SMILES:
CCOC(=O)NC(=O)OCC
1
–
1
of
1
results
1
Promotions Available
Diethyl iminodicarboxylate, 98%
Diethyl iminodicarboxylate, 98%
CAS: 19617-44-8 Molecular Formula: C6H11NO4 Molecular Weight (g/mol): 161.16 InChI Key: PQVSTLUFSYVLTO-UHFFFAOYSA-N Synonym: azamalonic ester,diethyl imidodicarbonate,diethyl azamalonate,imidodicarbonic acid, diethyl ester,diethyl iminodicarboxylate,diethylazamalonate,imidodicarbonic acid,1,3-diethyl ester,diethylimidodicarbonate,diethyl imidodicarbonate #,ethyl ethoxycarbonylamino formate PubChem CID: 140564 IUPAC Name: ethyl N-ethoxycarbonylcarbamate SMILES: CCOC(=O)NC(=O)OCC