accessibility menu, dialog, popup

UserName

CAS RN 19594-40-2

H3COCH3CH3

CAS RN 19594-40-2

IUPAC Name: 2-methylundecan-4-one
Synonyms: 2-methylundecan-4-one
Molecular Weight (g/mol): 184.32
Molecular Formula: C12H24O
InChi Key: IEBAJFDSHJYDCK-UHFFFAOYSA-N
SMILES: CCCCCCCC(=O)CC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results