accessibility menu, dialog, popup

UserName

CAS RN 19535-47-8

NNNN

CAS RN 19535-47-8

IUPAC Name: quinoxalino[2,3-f]1,10-phenanthroline
Synonyms: dipyridophenazine dppz
Molecular Weight (g/mol): 282.31
Molecular Formula: C18H10N4
InChi Key: BVQAWSJMUYMNQN-UHFFFAOYSA-N
SMILES: C1=CC2=NC3=C(N=C2C=C1)C1=CC=CN=C1C1=NC=CC=C31
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results