accessibility menu, dialog, popup

UserName

CAS RN 19523-57-0

H3COO

CAS RN 19523-57-0

IUPAC Name: 2-ethoxynaphthalene-1-carbaldehyde
Synonyms: 2-ethoxynaphthalene-1-carbaldehyde
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: IMNKQTWVJHODOS-UHFFFAOYSA-N
SMILES: CCOC1=CC=C2C=CC=CC2=C1C=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results