accessibility menu, dialog, popup

UserName

CAS RN 1948-33-0

IUPAC Name: 2-tert-butylbenzene-1,4-diol
Synonyms: tert-butylhydroquinone TBHQ
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: BGNXCDMCOKJUMV-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(O)=CC=C1O
Boiling Point 
  • (2)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (2)
  • (7)
  • (2)
  • (3)
  • (4)
  • (2)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)