accessibility menu, dialog, popup

UserName

CH3NH3CH3CH3CONOO

CAS RN 1941-26-0

IUPAC Name: tetraethylazanium nitrate
Synonyms: tetraethylammonium nitrate
Molecular Weight (g/mol): 192.26
Molecular Formula: C8H20N2O3
InChi Key: JTJKNAJRGLQKDZ-UHFFFAOYSA-N
SMILES: [O-][N+]([O-])=O.CC[N+](CC)(CC)CC
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)