accessibility menu, dialog, popup

UserName

CAS RN 19311-91-2

H3CNH3COHO

CAS RN 19311-91-2

IUPAC Name: N,N-diethyl-2-hydroxybenzamide
Synonyms: salicylamide, N,N-diethyl-
Molecular Weight (g/mol): 193.25
Molecular Formula: C11H15NO2
InChi Key: ZVYXEXAXXWINEH-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=CC=C1O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results