accessibility menu, dialog, popup

UserName

CAS RN 193086-74-7

HNOOOHOFCH3

CAS RN 193086-74-7

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-fluorophenyl)-2-methylpropanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(2-fluorophenyl)-2-methylpropanoic acid
Molecular Weight (g/mol): 419.45
Molecular Formula: C25H22FNO4
InChi Key: VNYGHKCAXQKZEQ-RUZDIDTESA-N
SMILES: CC(CC1=C(F)C=CC=C1)(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)