accessibility menu, dialog, popup

UserName

CAS RN 192642-85-6

IUPAC Name: 2-(5-bromopyridin-2-yl)acetic acid
Synonyms: (5-bromopyridin-2-yl)acetic acid
Molecular Weight (g/mol): 216.03
Molecular Formula: C7H6BrNO2
InChi Key: ATKULCGQSLCGEK-UHFFFAOYSA-N
SMILES: OC(=O)CC1=NC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results