accessibility menu, dialog, popup

UserName

H3CNCH3H3CH3COClOOO

CAS RN 1923-70-2

IUPAC Name: tetrabutylazanium perchlorate
Synonyms: tetrabutylammonium ion perchlorate
Molecular Weight (g/mol): 341.92
Molecular Formula: C16H36ClNO4
InChi Key: KBLZDCFTQSIIOH-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (11)
Quantity 
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (6)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (2)
Grade 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (8)
  • (2)
  • (11)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)