accessibility menu, dialog, popup

UserName

BOH3COCH3HOOH

CAS RN 192182-54-0

IUPAC Name: (3,5-dimethoxyphenyl)boronic acid
Synonyms: 3,5-dimethoxyphenylboronic acid
Molecular Weight (g/mol): 181.98
Molecular Formula: C8H11BO4
InChi Key: XUIURRYWQBBCCK-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1)B(O)O
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results