accessibility menu, dialog, popup

UserName

CAS RN 1918-00-9

IUPAC Name: 3,6-dichloro-2-methoxybenzoic acid
Synonyms: dicamba
Molecular Weight (g/mol): 221.03
Molecular Formula: C8H6Cl2O3
InChi Key: IWEDIXLBFLAXBO-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(Cl)=C1C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results