missing translation for 'onlineSavingsMsg'
Learn More

Dicamba, Restek

Catalog No. 06719872
Encompass
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06719872 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06719872 Supplier Chromatographic Supplier No. RK 32247

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1918-00-9
Molecular Formula C8H6Cl2O3
Molecular Weight (g/mol) 221.033
InChI Key IWEDIXLBFLAXBO-UHFFFAOYSA-N
PubChem CID 3030
ChEBI CHEBI:81856
IUPAC Name 3,6-dichloro-2-methoxybenzoic acid
SMILES COC1=C(C=CC(=C1C(=O)O)Cl)Cl

Specifications

Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1000ug/mL, Methanol
Chemical Name or Material Dicamba, Herbicide Mix