accessibility menu, dialog, popup

UserName

CAS RN 190601-12-8

CH3H3CONCCHNOCH3H3CHCHCFeFe

CAS RN 190601-12-8

IUPAC Name: bis(λ²-iron(2+)) 2',5-bis[4-(propan-2-yl)-4,5-dihydro-1,3-oxazol-2-yl]-[1,1'-bi(cyclopentane)]-1(5),1'(5'),3,3'-tetraene-2,2'-diide bis(cyclopenta-2,4-dien-1-ide)
Synonyms: bis(λ²-iron(2+)) 2',5-bis(4-isopropyl-4,5-dihydro-1,3-oxazol-2-yl)-[1,1'-bi(cyclopentane)]-1(5),1'(5'),3,3'-tetraene-2,2'-diide bis(cyclopentadienide)
Molecular Weight (g/mol): 592.34
Molecular Formula: C32H36Fe2N2O2
InChi Key: GSVKKVGUOIENJR-UHFFFAOYNA-N
SMILES: [Fe++].[Fe++].[CH-]1C=CC=C1.[CH-]1C=CC=C1.CC(C)C1COC(=N1)[C-]1C=CC=C1C1=C(C=C[CH-]1)C1=NC(CO1)C(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results