accessibility menu, dialog, popup

UserName

H3COOH

CAS RN 18979-72-1

IUPAC Name: 3-butoxyphenol
Synonyms: M-butoxyphenol
Molecular Weight (g/mol): 166.22
Molecular Formula: C10H14O2
InChi Key: VGIJZDWQVCXVNL-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=CC(O)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results